Assay Design



Gene Transcripts

Gene Symbol
PRAC2
Entrez Gene ID
102133474
Gene Name
PRAC2 small nuclear protein
Gene Aliases
-
Chromosome Location
Chr.16: 34074836 - 34077144 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005583579.2 XP_005583636.1 1 - 2 1358 102

View Full Product Details