Assay Design



Gene Transcripts

Gene Symbol
KRT16
Entrez Gene ID
102134405
Gene Name
keratin 16
Gene Aliases
-
Chromosome Location
Chr.16: 53268778 - 53271899 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015438486.1 XP_015293972.1 6 - 7 1480 90

View Full Product Details