Assay Design



Gene Transcripts

Gene Symbol
TSR2
Entrez Gene ID
102133281
Gene Name
TSR2 ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.X: 53547949 - 53553089 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005593678.2 XP_005593735.1 4 - 5 481 101
XM_015443497.1 XP_015298983.1 4 - 5 498 101

View Full Product Details