Assay Design



Gene Transcripts

Gene Symbol
LOC102141865
Entrez Gene ID
102141865
Gene Name
pepsin A-4
Gene Aliases
PGA4
Chromosome Location
Chr.14: 13024202 - 13033017 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577682.1 XP_005577739.1 8 - 9 1207 82

View Full Product Details