Assay Design



Gene Transcripts

Gene Symbol
LOC102125111
Entrez Gene ID
102125111
Gene Name
platelet factor 4
Gene Aliases
PF4
Chromosome Location
Chr.5: 60588157 - 60589436 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555082.2 XP_005555139.1 1 - 2 252 87

View Full Product Details