Assay Design



Gene Transcripts

Gene Symbol
CYP4F12
Entrez Gene ID
102131739
Gene Name
cytochrome P450 family 4 subfamily F member 12
Gene Aliases
-
Chromosome Location
Chr.19: 15994409 - 16016012 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283264.1 NP_001270193.1 1 - 2 249 117
XM_015440166.1 XP_015295652.1 2 - 3 310 117
GenBank mRNA DQ074801.1 - 1 - 2 249 117

View Full Product Details