Assay Design



Gene Transcripts

Gene Symbol
TNFSF14
Entrez Gene ID
102130835
Gene Name
TNF superfamily member 14
Gene Aliases
-
Chromosome Location
Chr.19: 6851561 - 6856690 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287647.1 NP_001274576.1 3 - 4 303 70
GenBank mRNA HQ717157.1 - 3 - 4 303 70

View Full Product Details