Product Details
- Assay Reference Genome
Location
Chr.8:24576282 on build GRCm38
- Cytoband
- 8A2
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 209176
- Gene Symbol
- Ido2
- Gene Name
- indoleamine 2,3-dioxygenase 2
- Gene Aliases
- AI265623, C230043N17Rik, Ido-2, Indol1
- Location
- Chr.8:24531892-24576404 on build GRCm38
- Assay Gene Location
- Within Exon 1
View Full Product Details