Product Details

Assay Reference Genome
Location

Chr.9:59560942 on build GRCm38
Cytoband
9B
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
15211
Gene Symbol
Hexa
Gene Name
hexosaminidase A
Gene Aliases
Hex-1
Location
Chr.9:59539545-59565109 on build GRCm38
Assay Gene Location
Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Hexa NM_010421.4 8 908 NP_034551.2
AK075895.1 8 884 BAC36036.1
AK075911.1 8 909 BAC36049.1
AK080777.1 8 881 BAC38018.1
AK144168.1 8 1036 BAE25744.1
AK150444.1 8 879 BAE29566.1
AK151960.1 8 883 BAE30831.1
AK159091.1 9 879 BAE34808.1
AK159814.1 8 894 BAE35394.1
AK159890.1 8 881 BAE35457.1
AK160112.1 8 868 BAE35636.1
AK167013.1 8 894 BAE39189.1
AK168186.1 8 908 BAE40146.1
AK170903.1 8 895
AK179213.1
AK218113.1 3 255
BC010755.1 8 877 AAH10755.1
U07631.1 8 900 AAA18775.1
X64331.1 8 851 CAA45615.1

View Full Product Details