Product Details

Assay Reference Genome
Location

Chr.10:21142025 on build GRCm38
Cytoband
10A3
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
17863
Gene Symbol
Myb
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Location
Chr.10:21124930-21160984 on build GRCm38
Assay Gene Location
Within Exon 12
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Myb NM_001198914.1 12 2019 NP_001185843.1
NM_010848.3 11 1662 NP_034978.3
AK088079.1 12 1940 BAC40133.1
AK132250.1 11 1527
AK137794.1 5 607 BAE23498.1
AK137965.1 11 1528
BC011513.1 11 1570 AAH11513.1
M12848.1 11 1660 AAB59713.1
S83053.1 2 193 AAN86760.1
X02774.1 11 1432 CAA26551.1

View Full Product Details