Product Details

Assay Reference Genome
Location

Chr.10:21148779 on build GRCm38
Cytoband
10A3
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
17863
Gene Symbol
Myb
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Location
Chr.10:21124930-21160984 on build GRCm38
Assay Gene Location
Overlaps Intron 7 - Exon 7
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Myb NM_001198914.1 NP_001185843.1
NM_010848.3 NP_034978.3
AK036518.1 1 370
AK088079.1 BAC40133.1
AK088598.1 BAC40443.1
AK132250.1
AK137794.1 BAE23498.1
AK137965.1
BC011513.1 AAH11513.1
M12848.1 AAB59713.1
X02774.1 CAA26551.1

View Full Product Details