Product Details
- Assay Reference Genome
Location
Chr.11:110069516 on build GRCm38
- Cytoband
- 11E1
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 217258
- Gene Symbol
- Abca8a
- Gene Name
- ATP-binding cassette, sub-family A (ABC1), member 8a
- Gene Aliases
- Location
- Chr.11:110025634-110097583 on build GRCm38
- Assay Gene Location
- Overlaps Intron 14 - Exon 15
View Full Product Details