Product Details

Assay Reference Genome
Location

Chr.11:110030380 on build GRCm38
Cytoband
11E1
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
217258
Gene Symbol
Abca8a
Gene Name
ATP-binding cassette, sub-family A (ABC1), member 8a
Gene Aliases
Location
Chr.11:110025634-110097583 on build GRCm38
Assay Gene Location
Overlaps Intron 34 - Exon 35
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Abca8a NM_153145.4 NP_694785.3
AB255881.1
AB255882.1
AF498360.1 AAM90906.1
AK031843.1 BAC27576.1
AY732492.1 AAU81985.1
BC026496.1 AAH26496.1
BC060032.1 AAH60032.1

View Full Product Details