Assay Design
Gene Transcripts
- Gene Symbol
- Alk
- Entrez Gene ID
-
11682
- Gene Name
- anaplastic lymphoma kinase
- Gene Aliases
-
CD246, Tcrz
- Chromosome Location
-
Chr.17: 71868988 - 72605410 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_007439.2 |
NP_031465.2 |
15 - 16 |
3239 |
70 |
View Full Product Details