Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007572.2 | NP_031598.2 | 2 - 3 | 271 | 80 |
GenBank mRNA | AK002655.1 | - | 2 - 3 | 197 | 80 |
AK136737.1 | - | 2 - 3 | 197 | 80 | |
AK150771.1 | - | 2 - 3 | 197 | 80 | |
AK159210.1 | - | 2 - 3 | 197 | 80 | |
AK159343.1 | - | 2 - 3 | 192 | 80 | |
BC002086.1 | - | 2 - 3 | 230 | 80 | |
BC020096.1 | - | 1 - 2 | 193 | 80 | |
BY318890.1 | - | 2 - 3 | 274 | 80 | |
X58861.1 | - | 1 - 2 | 194 | 80 |