Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007576.3 | NP_031602.3 | 5 - 6 | 724 | 78 |
GenBank mRNA | AB094485.1 | - | - | 705 | 78 |
AB094486.1 | - | 5 - 6 | 640 | 78 | |
AK136735.1 | - | 5 - 6 | 638 | 78 | |
AK149468.1 | - | 5 - 6 | 731 | 78 | |
AK149477.1 | - | 5 - 6 | 641 | 78 | |
AK162345.1 | - | 5 - 6 | 638 | 78 | |
BC012257.1 | - | 5 - 6 | 696 | 78 | |
M17122.1 | - | 5 - 6 | 738 | 78 |