Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044741.2 | NP_001038206.1 | 2 - 3 | 320 | 85 |
NM_001286226.1 | NP_001273155.1 | 2 - 3 | 574 | 85 | |
NM_007581.3 | NP_031607.2 | 2 - 3 | 574 | 85 | |
GenBank mRNA | AF498245.1 | - | 2 - 3 | 172 | 85 |
AF498246.1 | - | 2 - 3 | 173 | 85 | |
AK148088.1 | - | 2 - 3 | 575 | 85 | |
AK197162.1 | - | 1 - 2 | 52 | 85 | |
BC055926.1 | - | 1 - 2 | 126 | 85 | |
BU709207.1 | - | 2 - 3 | 165 | 85 | |
CD807951.1 | - | 2 - 3 | 428 | 85 | |
CJ131971.1 | - | 2 - 3 | 313 | 85 | |
U20372.1 | - | 2 - 3 | 228 | 85 |