Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007661.3 | NP_031687.2 | 7 - 8 | 839 | 62 |
GenBank mRNA | AK008283.1 | - | 7 - 8 | 816 | 62 |
AK077668.1 | - | 7 - 8 | 840 | 62 | |
AK135402.1 | - | 7 - 8 | 846 | 62 | |
AK147133.1 | - | 7 - 8 | 817 | 62 | |
BC052920.1 | - | 7 - 8 | 917 | 62 | |
L37092.1 | - | 6 - 7 | 1414 | 62 | |
M58633.1 | - | 2 - 3 | 63 |