Assay Design



Gene Transcripts

Gene Symbol
Crp
Entrez Gene ID
12944
Gene Name
C-reactive protein, pentraxin-related
Gene Aliases
AI255847
Chromosome Location
Chr.1: 172698056 - 172699966 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007768.4 NP_031794.3 1 - 2 136 134
GenBank mRNA AK088477.1 - 1 - 2 126 134
BC011124.1 - 1 - 2 126 134
X17496.1 - 1 - 2 136 134

View Full Product Details