Assay Design



Gene Transcripts

Gene Symbol
Dvl3
Entrez Gene ID
13544
Gene Name
dishevelled 3, dsh homolog (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.16: 20516938 - 20532548 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007889.2 NP_031915.2 1 - 2 295 75
GenBank mRNA AK052151.1 - 1 - 2 296 75
BC033414.1 - 1 - 2 173 75
U41285.1 - 1 - 2 377 75

View Full Product Details