Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161373.1 | NP_001154845.1 | 19 - 20 | 6287 | 73 |
NM_001161374.1 | NP_001154846.1 | 19 - 20 | 6287 | 73 | |
NM_007977.2 | NP_032003.2 | 20 - 21 | 6497 | 73 | |
GenBank mRNA | BC138347.1 | - | 19 - 20 | 6168 | 73 |
BC145245.1 | - | 19 - 20 | 6168 | 73 | |
L05573.1 | - | 20 - 21 | 6495 | 73 |