Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008004.4 | NP_032030.1 | 2 - 3 | 583 | 84 |
GenBank mRNA | AB009250.1 | - | 2 - 3 | 87 | 84 |
AK077555.1 | - | 2 - 3 | 585 | 84 | |
AK082858.1 | - | 2 - 3 | 474 | 84 | |
BC119033.1 | - | 2 - 3 | 569 | 84 |