Assay Design
Gene Transcripts
- Gene Symbol
- Il9
- Entrez Gene ID
-
16198
- Gene Name
- interleukin 9
- Gene Aliases
-
Il-9, P40
- Chromosome Location
-
Chr.13: 56479277 - 56482246 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_008373.1 |
NP_032399.1 |
2 - 3 |
163 |
64 |
GenBank mRNA |
X14045.1 |
- |
2 - 3 |
163 |
64 |
View Full Product Details