Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008404.4 | NP_032430.2 | 6 - 7 | 860 | 65 |
GenBank mRNA | AK088435.1 | - | 6 - 7 | 849 | 65 |
AK136502.1 | - | 6 - 7 | 861 | 65 | |
AK161236.1 | - | 6 - 7 | 829 | 65 | |
AK169587.1 | - | 6 - 7 | 895 | 65 | |
BC145644.1 | - | 5 - 6 | 782 | 65 | |
M31039.1 | - | 5 - 6 | 748 | 65 | |
X14951.1 | - | 6 - 7 | 826 | 65 |