Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008501.2 | NP_032527.1 | 2 - 3 | 243 | 64 |
GenBank mRNA | AF065917.1 | - | 1 - 2 | 35 | 64 |
AF065918.1 | - | 1 - 2 | 35 | 64 | |
BY207460.1 | - | 2 - 3 | 244 | 64 | |
BY209921.1 | - | 1 - 2 | 85 | 64 | |
X12810.1 | - | 1 - 2 | 35 | 64 |