Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113198.1 | NP_001106669.1 | 8 - 9 | 1159 | 68 |
NM_001178049.1 | NP_001171520.1 | 8 - 9 | 1046 | 68 | |
NM_008601.3 | NP_032627.1 | 7 - 8 | 850 | 68 | |
GenBank mRNA | AB221657.1 | - | 7 - 8 | 712 | 68 |
AK052895.1 | - | 8 - 9 | 1018 | 68 | |
AK155350.1 | - | 8 - 9 | 1160 | 68 | |
BC108976.2 | - | 7 - 8 | 814 | 68 | |
BC108977.1 | - | 7 - 8 | 779 | 68 | |
Z23066.1 | - | 7 - 8 | 841 | 68 |