Assay Design



Gene Transcripts

Gene Symbol
Myf5
Entrez Gene ID
17877
Gene Name
myogenic factor 5
Gene Aliases
B130010J22Rik, Myf-5, bHLHc2
Chromosome Location
Chr.10: 107482908 - 107486927 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008656.5 NP_032682.1 1 - 2 702 58
GenBank mRNA AF336978.1 - 1 - 2 223 58
AK044894.1 - 1 - 2 656 58
AK083402.1 - 1 - 2 704 58
BC132144.1 - 1 - 2 556 58
X56182.1 - 1 - 2 499 58

View Full Product Details