Assay Design



Gene Transcripts

Gene Symbol
Abcb4
Entrez Gene ID
18670
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 4
Gene Aliases
Mdr2, Pgy-2, Pgy2, mdr-2
Chromosome Location
Chr.5: 8893598 - 8959226 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008830.2 NP_032856.2 1 - 2 189 56
GenBank mRNA BC150687.1 - 1 - 2 59 56
J03398.1 - 1 - 2 189 56

View Full Product Details