Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008871.2 | NP_032897.2 | 5 - 6 | 1044 | 87 |
GenBank mRNA | BC054091.1 | - | 5 - 6 | 1041 | 87 |
GQ872458.1 | - | 4 - 5 | 899 | 87 | |
GQ872459.1 | - | 4 - 5 | 899 | 87 | |
M33960.1 | - | 5 - 6 | 1030 | 87 |