Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009311.2 | NP_033337.1 | 1 - 2 | 94 | 77 |
GenBank mRNA | BC117081.1 | - | 1 - 2 | 69 | 77 |
BC119122.1 | - | 1 - 2 | 69 | 77 | |
BC144737.1 | - | 1 - 2 | 69 | 77 | |
D17584.1 | - | 1 - 2 | 94 | 77 |