Assay Design



Gene Transcripts

Gene Symbol
Tacr2
Entrez Gene ID
21337
Gene Name
tachykinin receptor 2
Gene Aliases
Nk2r, Skr, Tac2r
Chromosome Location
Chr.10: 62252438 - 62265990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009314.4 NP_033340.3 1 - 2 771 71
GenBank mRNA AK033443.1 - 1 - 2 454 71
BC106959.1 - 1 - 2 420 71
BC106960.2 - 4 - 5 71
X62933.1 - 1 - 2 771 71

View Full Product Details