Assay Design



Gene Transcripts

Gene Symbol
Tial1
Entrez Gene ID
21843
Gene Name
Tia1 cytotoxic granule-associated RNA binding protein-like 1
Gene Aliases
5330433G13Rik, AL033329, TIAR, mTIAR
Chromosome Location
Chr.7: 128439772 - 128461668 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009383.2 NP_033409.1 1 - 2 387 74
GenBank mRNA BC010496.1 - 2 - 3 172 74
BC056227.1 - 1 - 2 83 74
U55861.1 - 1 - 2 163 74

View Full Product Details