Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083927.1 | NP_001077396.1 | 10 - 11 | 1871 | 88 |
NM_001083928.1 | NP_001077397.1 | 10 - 11 | 1841 | 88 | |
NM_009389.2 | NP_033415.2 | 10 - 11 | 1841 | 88 | |
GenBank mRNA | AK031322.1 | - | 8 - 9 | 648 | 88 |
AK122524.1 | - | 10 - 11 | 1349 | 88 | |
AK147635.1 | - | 2 - 3 | 5068 | 88 | |
AK158785.1 | - | 9 - 10 | 1161 | 88 | |
BC056465.1 | - | 10 - 11 | 852 | 88 | |
X73360.1 | - | 9 - 10 | 1024 | 88 |