Assay Design



Gene Transcripts

Gene Symbol
Tnfrsf11a
Entrez Gene ID
21934
Gene Name
tumor necrosis factor receptor superfamily, member 11a, NFKB activator
Gene Aliases
Ly109, ODFR, OFE, Rank, TRANCE-R, mRANK
Chromosome Location
Chr.1: 105780723 - 105847981 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009399.3 NP_033425.3 8 - 9 817 73
GenBank mRNA AF019046.1 - 8 - 9 812 73
AK138990.1 - 8 - 9 823 73
BC019185.1 - 8 - 9 814 73

View Full Product Details