Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009469.3 | NP_033495.2 | 27 - 28 | 3457 | 66 |
GenBank mRNA | AF053756.1 | - | 27 - 28 | 3149 | 66 |
AF072370.1 | - | 27 - 28 | 3140 | 66 | |
AK122357.1 | - | 26 - 27 | 2959 | 66 | |
AK148317.1 | - | 27 - 28 | 3450 | 66 | |
AK154939.1 | - | 27 - 28 | 3474 | 66 | |
AK156534.1 | - | 27 - 28 | 3434 | 66 | |
BC023281.1 | - | 3 - 4 | 383 | 66 | |
BC030850.1 | - | 6 - 7 | 929 | 66 | |
BC053062.1 | - | 27 - 28 | 3117 | 66 | |
BC057121.1 | - | 27 - 28 | 3457 | 66 | |
BC059835.1 | - | 27 - 28 | 3426 | 66 |