Product Details

Assay Reference Genome
Location

Chr.15:91712804 on build GRCm38
Cytoband
15E3
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
66725
Gene Symbol
Lrrk2
Gene Name
leucine-rich repeat kinase 2
Gene Aliases
4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
Location
Chr.15:91672821-91816124 on build GRCm38
Assay Gene Location
Within Exon 14
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Lrrk2 NM_025730.3 14 1655 NP_080006.3
AK052591.1 14 1656 BAC35052.1
AY792512.1 14 1598 AAV63976.1

View Full Product Details