Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256224.1 | NP_001243153.1 | 2 - 3 | 153 | 98 |
NM_009524.3 | NP_033550.2 | 2 - 3 | 776 | 98 | |
GenBank mRNA | AK031512.1 | - | 2 - 3 | 486 | 98 |
AK031597.1 | - | 2 - 3 | 749 | 98 | |
AK036824.1 | - | 2 - 3 | 154 | 98 | |
BC018425.1 | - | 2 - 3 | 393 | 98 | |
M89798.1 | - | 2 - 3 | 690 | 98 |