Assay Design



Gene Transcripts

Gene Symbol
Wnt6
Entrez Gene ID
22420
Gene Name
wingless-type MMTV integration site family, member 6
Gene Aliases
AA409270, Wnt-6
Chromosome Location
Chr.1: 74754344 - 74785322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009526.3 NP_033552.2 1 - 2 335 87
GenBank mRNA BC048700.1 - 1 - 2 269 87
M89800.1 - 1 - 2 81 87

View Full Product Details