Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009724.2 | NP_033854.1 | 6 - 7 | 755 | 125 |
GenBank mRNA | AK008180.1 | - | 6 - 7 | 754 | 125 |
AK009499.1 | - | 6 - 7 | 755 | 125 | |
AK009994.1 | - | 6 - 7 | 754 | 125 | |
AK010003.1 | - | 6 - 7 | 754 | 125 | |
AK010049.1 | - | 6 - 7 | 802 | 125 | |
BC120705.1 | - | 6 - 7 | 754 | 125 | |
BC125292.1 | - | 6 - 7 | 754 | 125 |