Product Details
- Assay Reference Genome
Location
Chr.15:91737204 on build GRCm38
- Cytoband
- 15E3
- Species
- Mus musculus
- Variation Type
- Copy Number
Additional Information
Target Gene Details
- Entrez Gene ID
- 66725
- Gene Symbol
- Lrrk2
- Gene Name
- leucine-rich repeat kinase 2
- Gene Aliases
- 4921513O20Rik, 9330188B09Rik, AW561911, D630001M17Rik, Gm927, cI-46
- Location
- Chr.15:91672821-91816124 on build GRCm38
- Assay Gene Location
- Within Exon 23
View Full Product Details