Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009787.2 | NP_033917.2 | 4 - 5 | 812 | 86 |
GenBank mRNA | AK028292.1 | - | 4 - 5 | 662 | 86 |
AK146288.1 | - | 4 - 5 | 645 | 86 | |
AK148801.1 | - | 4 - 5 | 732 | 86 | |
AK150566.1 | - | 4 - 5 | 693 | 86 | |
AK161534.1 | - | 4 - 5 | 650 | 86 | |
AK169387.1 | - | 4 - 5 | 657 | 86 | |
BC066857.1 | - | 3 - 4 | 411 | 86 | |
BC141078.1 | - | 4 - 5 | 708 | 86 | |
J05186.1 | - | 4 - 5 | 695 | 86 | |
Y00884.1 | - | 3 - 4 | 518 | 86 |