Assay Design



Gene Transcripts

Gene Symbol
En1
Entrez Gene ID
13798
Gene Name
engrailed 1
Gene Aliases
En-1, Mo-en.1
Chromosome Location
Chr.1: 120602391 - 120607991 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010133.2 NP_034263.2 1 - 2 1433 90
GenBank mRNA AK140408.1 - 1 - 2 1296 90
L12703.1 - 1 - 2 1239 90

View Full Product Details