Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010166.3 | NP_034296.2 | 8 - 9 | 663 | 103 |
NM_210071.2 | NP_997592.1 | 7 - 8 | 624 | 103 | |
NM_211357.2 | NP_997596.1 | 5 - 6 | 443 | 103 | |
GenBank mRNA | AK144653.1 | - | 7 - 8 | 615 | 103 |
AK164591.1 | - | 7 - 8 | 624 | 103 | |
BC052860.1 | - | 6 - 7 | 427 | 103 | |
BC063259.1 | - | 7 - 8 | 552 | 103 | |
BY730487.1 | - | 7 - 8 | 624 | 103 | |
U61112.1 | - | 4 - 5 | 328 | 103 | |
U81604.1 | - | 8 - 9 | 631 | 103 |