Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010252.4 | NP_034382.2 | 8 - 9 | 1527 | 70 |
GenBank mRNA | AF156490.1 | - | 1 - 2 | 92 | 70 |
AK032128.1 | - | 8 - 9 | 1212 | 70 | |
AK034085.1 | - | 8 - 9 | 992 | 70 | |
AK034332.1 | - | 8 - 9 | 1528 | 70 | |
AK038468.1 | - | 8 - 9 | 1264 | 70 | |
AK047752.1 | - | 8 - 9 | 1277 | 70 | |
AK049956.1 | - | 8 - 9 | 1265 | 70 | |
AK162884.1 | - | 9 - 10 | 1210 | 70 | |
AK163541.1 | - | 8 - 9 | 1236 | 70 | |
BC099939.1 | - | 8 - 9 | 1236 | 70 | |
X55272.1 | - | 8 - 9 | 1310 | 70 |