Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010424.4 | NP_034554.2 | 3 - 4 | 745 | 83 |
GenBank mRNA | AK009581.1 | - | 3 - 4 | 746 | 83 |
AK088986.1 | - | 3 - 4 | 741 | 83 | |
AK150697.1 | - | 3 - 4 | 763 | 83 | |
AK220051.1 | - | 1 - 2 | 59 | 83 | |
BC116744.1 | - | 3 - 4 | 703 | 83 | |
BC116746.1 | - | 3 - 4 | 701 | 83 | |
U66849.1 | - | 3 - 4 | 727 | 83 |