Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010552.3 | NP_034682.1 | 2 - 3 | 296 | 91 |
GenBank mRNA | AK040420.1 | - | 2 - 3 | 297 | 91 |
BC119303.1 | - | 2 - 3 | 296 | 91 | |
BC119309.1 | - | 2 - 3 | 296 | 91 | |
U43088.1 | - | 2 - 3 | 283 | 91 |