Assay Design



Gene Transcripts

Gene Symbol
Insr
Entrez Gene ID
16337
Gene Name
insulin receptor
Gene Aliases
4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-B
Chromosome Location
Chr.8: 3150922 - 3279649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010568.2 NP_034698.2 4 - 5 1617 79
GenBank mRNA J05149.1 - 4 - 5 1176 79
KC356871.1 - 4 - 5 1128 79

View Full Product Details