Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010766.2 | NP_034896.1 | 11 - 12 | 1307 | 69 |
GenBank mRNA | AK156388.1 | - | 11 - 12 | 981 | 69 |
AK159967.1 | - | 11 - 12 | 989 | 69 | |
AK169230.1 | - | 11 - 12 | 983 | 69 | |
BC132338.1 | - | 11 - 12 | 1247 | 69 | |
BC137939.1 | - | 11 - 12 | 1247 | 69 | |
U18424.1 | - | 11 - 12 | 1159 | 69 |