Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285853.1 | NP_001272782.1 | 9 - 10 | 1134 | 63 |
NM_001285854.1 | NP_001272783.1 | 9 - 10 | 1131 | 63 | |
NM_001285855.1 | NP_001272784.1 | 9 - 10 | 1199 | 63 | |
NM_010768.2 | NP_034898.1 | 10 - 11 | 1267 | 63 | |
GenBank mRNA | AK013606.1 | - | 9 - 10 | 948 | 63 |
D45243.1 | - | 9 - 10 | 1199 | 63 | |
L27738.1 | - | 10 - 11 | 1021 | 63 | |
S77473.1 | - | 9 - 10 | 946 | 63 | |
U05210.1 | - | 9 - 10 | 920 | 63 |