Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011035.2 | NP_035165.2 | 14 - 15 | 1777 | 114 |
GenBank mRNA | AF082077.1 | - | 14 - 15 | 1742 | 114 |
AK077497.1 | - | 14 - 15 | 1742 | 114 | |
BC055740.1 | - | 11 - 12 | 1425 | 114 | |
BC060157.1 | - | 11 - 12 | 1425 | 114 | |
BC079838.1 | - | 14 - 15 | 1762 | 114 |